missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ 8 Cal UltraSoft Khaki Coverall 30 in. Inseam
Description
- Arc Rating: 8 cal/cm²
- CAT 2
- Fabric/Material: 7 oz. Westex™ UltraSoft™
- Action back allows for full range of motion
- All metal covered to ensure no contact with skin
- Chest pockets with flaps and hidden snap closures
- Comfortable sewn-in elastic waist for better fit
- Convenient side pass-through pockets for access to pant pockets
- Covered zipper front for easy entry
- FR fabric will not melt, drip or ignite
- FR protection from electrical arc flash and flash fire hazards
- Home laundry
- Leg openings made to fit over work boots
- Two front pockets with pass-through for access to pants pockets
- Made in the USA
Specifications
Specifications
| Size | Small |
| Material | Cotton, Nylon |
| Color | Khaki |
| Ankle Style | Open |
| Closure Type | Zipper |
| Seam Type | Stitched |
| Certifications/Compliance | Arc Rating: 8 cal/cm², CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269 |
| Suspender | None |