missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ 8 Cal UltraSoft FR Coveralls 30 in. Inseam

Coveralls with comfortable sewn-in elastic waist for better fit

Supplier:  National Safety Apparel™ C88UJXL30

 View more versions of this product

Catalog No. 17001440

Only null left
Explore available promotions

Description

Description

  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Home laundry
  • Made in the USA
Specifications

Specifications

X-Large
Navy
Covered Zipper
Arc Rating: 8 cal/cm², CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Cotton, Nylon
Open
Stitched
None
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions