Learn More
National Safety Apparel™ 8 Cal UltraSoft Arc Flash Coat
Catalog No. p-7220070
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001221 Small
17001222 Medium
17001223 Large
17001224 2X-Large
17001225 3X-Large
17001226 X-Large
17001227 4X-Large
17001228 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001221 Supplier National Safety Apparel™ Supplier No. C04UJ03SM32
Only null left
For enhanced protection Westex™ UltraSoft is the ideal choice for superior comfort, improved safety, and above average durability.
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • FR fabric will not melt, drip or ignite
  • FR knit wrist cuffs keep sleeves in place
  • Generous fit for ease of movement
  • Inherently flame resistant protection will not wash/wear out
  • Stand up collar lined with anti-microbial, high performance fabric
  • Hook and loop front closure
  • Made in the USA
Specifications
Closure Type Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269
Show More Show Less