Learn More
National Safety Apparel™ 8 Cal ArcGuard™ Protera Arc Flash Kit with FR Coverall (10)
Catalog No. p-7220184
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
170011090 X-Large
1 options
Catalog No. 170011090 Supplier National Safety Apparel™ Supplier No. KIT2CVPR08XL10
Only null left
With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. DuPont™ Protera™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Color Khaki
Closure Type Zipper
Suspender None
Ankle Style Open
Material DuPont™ Protera™
Seam Type Stitched
Size X-Large