missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ ArcGuard™ 8 cal Coverall Arc Flash Kit, Balaclava/Voltage Gloves (10)

Catalog No. 17001830
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17001830 5X-Large
17001795 Small
17001800 Medium
17001805 Large
17001810 X-Large
17001815 2X-Large
17001820 3X-Large
17001825 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17001830 Supplier National Safety Apparel™ Supplier No. KIT2CV08B5X10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The premium choice for superior comfort, improved safety and above average durability

  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper

Specifications

Gender Unisex
Product Type Arc Flash Kit
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Certifications/Compliance CAT 2, NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Arc Flash Rating 8 cal/cm²
Closure Type Zipper
Color Navy
Material Cotton, Nylon
Seam Type Stitched
Size 5X-Large
Suspender None
Ankle Style Open
Show More Show Less