missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoisoquinoline 97.0+%, TCI America™

Catalog No. B41441G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41441G 1 g
B41445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41441G Supplier TCI America Supplier No. B41441G

Chemical Identifiers

CAS 63927-22-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD04973298
InChI Key DPRIHFQFWWCIGY-UHFFFAOYSA-N
PubChem CID 9859134
IUPAC Name 8-bromoisoquinoline
SMILES BrC1=C2C=NC=CC2=CC=C1

Specifications

Melting Point 78°C
Color Yellow
Quantity 1 g
Synonym 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879
Formula Weight 208.06
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromoisoquinoline