Learn More
8-Bromoisoquinoline, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 63927-22-0 |
|---|---|
| Molecular Formula | C9H6BrN |
| Molecular Weight (g/mol) | 208.06 |
| MDL Number | MFCD04973298 |
| InChI Key | DPRIHFQFWWCIGY-UHFFFAOYSA-N |
| Synonym | 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879 |
| PubChem CID | 9859134 |
| IUPAC Name | 8-bromoisoquinoline |
| SMILES | BrC1=C2C=NC=CC2=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC458760010
|
Thermo Scientific Chemicals
458760010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC458760050
|
Thermo Scientific Chemicals
458760050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 63927-22-0 | |
| 208.06 | |
| DPRIHFQFWWCIGY-UHFFFAOYSA-N | |
| 9859134 | |
| BrC1=C2C=NC=CC2=CC=C1 |
| C9H6BrN | |
| MFCD04973298 | |
| 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879 | |
| 8-bromoisoquinoline |
Specifications
| 63927-22-0 | |
| 96% min. (GC) | |
| C9H6BrN | |
| 1g | |
| DPRIHFQFWWCIGY-UHFFFAOYSA-N | |
| 8-bromoisoquinoline | |
| 9859134 | |
| 97% |
| Authentic | |
| Glass Bottle | |
| MFCD04973298 | |
| 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879 | |
| BrC1=C2C=NC=CC2=CC=C1 | |
| 208.06 | |
| 208.06 | |
| 8-Bromoisoquinoline |
Safety and Handling
GHS H Statement Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
RUO â Research Use Only