missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoisoquinoline, 97%

Catalog No. AC458760010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458760010 1 g
AC458760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458760010 Supplier Thermo Scientific Chemicals Supplier No. 458760010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63927-22-0 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63927-22-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD04973298
InChI Key DPRIHFQFWWCIGY-UHFFFAOYSA-N
Synonym 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879
PubChem CID 9859134
IUPAC Name 8-bromoisoquinoline
SMILES BrC1=C2C=NC=CC2=CC=C1

Specifications

Melting Point 78.0°C to 81.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 8-Bromoisoquinoline
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only