missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoguanosine hydrate, 97%

CAS: 332359-99-6 | C10H12BrN5O5 | 362.14 g/mol

$39.00 - $109.62

Chemical Identifiers

CAS 332359-99-6
Molecular Formula C10H12BrN5O5
Molecular Weight (g/mol) 362.14
MDL Number MFCD00150531
InChI Key ASUCSHXLTWZYBA-UMMCILCDSA-N
Synonym 8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate
PubChem CID 18530343
IUPAC Name 2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate
SMILES NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0299203
View Documents
Thermo Scientific Chemicals
L0299203
1 g
Each for $39.00
Only null left
 
AAL0299206
View Documents
Thermo Scientific Chemicals
L0299206
5 g
Each for $109.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

332359-99-6
362.14
ASUCSHXLTWZYBA-UMMCILCDSA-N
18530343
NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
C10H12BrN5O5
MFCD00150531
8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate
2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate

Specifications

332359-99-6
C10H12BrN5O5
1 g
8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate
−30° (c=2 in 1:1 DMSO/Water)
2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate
18530343
97%
>300°C
MFCD00150531
578725
ASUCSHXLTWZYBA-UMMCILCDSA-N
NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
362.14
362.14 (Anhydrous)
8-Bromoguanosine hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-677-6

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only