missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromoadenine
CAS: 6974-78-3 | C5H4BrN5 | 214.03 g/mol
Supplier: Thermo Scientific Chemicals J6558203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 8-Bromoadenine | |
| >250°C | |
| C5H4BrN5 | |
| 1 g | |
| FVXHPCVBOXMRJP-UHFFFAOYSA-N | |
| 8-bromo-7H-purin-6-amine | |
| 81457 | |
| Powder |
| 6974-78-3 | |
| Brown to White | |
| MFCD00082518 | |
| 8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo | |
| NC1=C2NC(Br)=NC2=NC=N1 | |
| 214.03 | |
| 214.03 |
Chemical Identifiers
| 6974-78-3 | |
| 214.03 | |
| FVXHPCVBOXMRJP-UHFFFAOYSA-N | |
| 81457 | |
| NC1=C2NC(Br)=NC2=NC=N1 |
| C5H4BrN5 | |
| MFCD00082518 | |
| 8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo | |
| 8-bromo-7H-purin-6-amine |
Safety and Handling
EINECSNumber : 230-225-1
RTECSNumber : UO7410000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only