missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenine

Catalog No. AAJ65582ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65582ME 500 mg
AAJ65582MC 100 mg
AAJ6558203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ65582ME Supplier Thermo Scientific Chemicals Supplier No. J65582ME

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6974-78-3 | C5H4BrN5 | 214.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6974-78-3
Molecular Formula C5H4BrN5
Molecular Weight (g/mol) 214.03
MDL Number MFCD00082518
InChI Key FVXHPCVBOXMRJP-UHFFFAOYSA-N
Synonym 8-bromoadenine, 8-bromo-9h-purin-6-amine, 6-amino-8-bromopurine, 8-br-adenine, 8-bromo-1h-adenine, 8-bromo-1h-purin-6-amine, 8-bromo-9h-purin-6-ylamine, 8-bromopurine-6-ylamine, chembl65747, 9h-purin-6-amine, 8-bromo
PubChem CID 81457
IUPAC Name 8-bromo-7H-purin-6-amine
SMILES NC1=C2NC(Br)=NC2=NC=N1

Specifications

Melting Point >250°C
Color Brown to White
Quantity 500 mg
Formula Weight 214.03
Physical Form Powder
Chemical Name or Material 8-Bromoadenine
EINECSNumber 230-225-1
RTECSNumber UO7410000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only