missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine 98.0+%, TCI America™

Catalog No. B4454200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4454200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4454200MG Supplier TCI America Supplier No. B4454200MG
Only null left

Chemical Identifiers

CAS 666816-98-4
Molecular Formula C10H9BrN4O2
Molecular Weight (g/mol) 297.112
MDL Number MFCD22126091
InChI Key HFZOBQSHTNNKFY-UHFFFAOYSA-N
Synonym 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
PubChem CID 21088927
IUPAC Name 8-bromo-7-but-2-ynyl-3-methylpurine-2,6-dione
SMILES CC#CCN1C2=C(N=C1Br)N(C(=O)NC2=O)C

Specifications

Melting Point 285°C
Color White
Quantity 200 mg
Formula Weight 297.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine