missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-2-chloro-6-fluoroquinazoline, 95%, Thermo Scientific Chemicals

Catalog No. p-7056910
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66128MD 250 mg
AAH6612803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66128MD Supplier Thermo Scientific Chemicals Supplier No. H66128MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 953039-63-9
Molecular Formula C8H3BrClFN2
Molecular Weight (g/mol) 261.48
MDL Number MFCD18382554
InChI Key XTEWVYPXKPYPRE-UHFFFAOYSA-N
Synonym 8-bromo-2-chloro-6-fluoro-quinazoline, quinazoline, 8-bromo-2-chloro-6-fluoro
PubChem CID 59199299
IUPAC Name 8-bromo-2-chloro-6-fluoroquinazoline
SMILES FC1=CC(Br)=C2N=C(Cl)N=CC2=C1

Specifications

Quantity 250 mg
Beilstein 15583920
Formula Weight 261.48
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 8-Bromo-2-chloro-6-fluoroquinazoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only