missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octanol 90.0+%, TCI America™

Catalog No. p-7119228
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17295G 5 g
B172925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17295G Supplier TCI America Supplier No. B17295G
Only null left

Chemical Identifiers

CAS 50816-19-8
Molecular Formula C8H17BrO
Molecular Weight (g/mol) 209.127
MDL Number MFCD00010388
InChI Key GMXIEASXPUEOTG-UHFFFAOYSA-N
Synonym 8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
PubChem CID 162607
IUPAC Name 8-bromooctan-1-ol
SMILES C(CCCCBr)CCCO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 209.13
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Bromo-1-octanol
TSCA Yes
Recommended Storage Refrigerator