missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octanol, 95%

Catalog No. p-7030092
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2762803 1 g
AAH2762806 5 g
AAH2762814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2762803 Supplier Thermo Scientific Chemicals Supplier No. H2762803

CAS: 50816-19-8 | C8H17BrO | 209.127 g/mol

It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).

Chemical Identifiers

CAS 50816-19-8
Molecular Formula C8H17BrO
Molecular Weight (g/mol) 209.127
MDL Number MFCD00010388
InChI Key GMXIEASXPUEOTG-UHFFFAOYSA-N
Synonym 8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
PubChem CID 162607
IUPAC Name 8-bromooctan-1-ol
SMILES C(CCCCBr)CCCO

Specifications

Density 1.22
Boiling Point 79°C to 80°C (0.07 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.481
Linear Formula Br(CH2)8OH
Quantity 1 g
Beilstein 1900837
Formula Weight 209.13
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-octanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-785-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only