missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Amino-2-methylquinoline 98.0+%, TCI America™
$104.38 - $313.16
Chemical Identifiers
| CAS | 18978-78-4 |
|---|---|
| Molecular Formula | C10H10N2 |
| Molecular Weight (g/mol) | 158.20 |
| MDL Number | MFCD00023998 |
| InChI Key | JHIAOWGCGNMQKA-UHFFFAOYSA-N |
| Synonym | 8-Aminoquinaldine |
| PubChem CID | 140457 |
| IUPAC Name | 2-methylquinolin-8-amine |
| SMILES | CC1=CC=C2C=CC=C(N)C2=N1 |
Chemical Identifiers
| 18978-78-4 | |
| 158.20 | |
| JHIAOWGCGNMQKA-UHFFFAOYSA-N | |
| 140457 | |
| CC1=CC=C2C=CC=C(N)C2=N1 |
| C10H10N2 | |
| MFCD00023998 | |
| 8-Aminoquinaldine | |
| 2-methylquinolin-8-amine |
Specifications
| 18978-78-4 | |
| Yellow | |
| MFCD00023998 | |
| 8-Aminoquinaldine | |
| CC1=CC=C2C=CC=C(N)C2=N1 | |
| 158.20 | |
| 158.20 | |
| Crystalline Powder |
| 56°C | |
| C10H10N2 | |
| 1 g | |
| JHIAOWGCGNMQKA-UHFFFAOYSA-N | |
| 2-methylquinolin-8-amine | |
| 140457 | |
| ≥98.0% (T) | |
| 8-Amino-2-methylquinoline |
Safety and Handling
TSCA : No