missing translation for 'onlineSavingsMsg'
Learn More

8-Amino-1-octanol 98.0+%, TCI America™

Catalog No. A15221G
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A15221G 1 g
A15225G 5 g
2 options

Catalog No. A15221G

Supplier: TCI America A15221G

Only null left

Chemical Identifiers

CAS 19008-71-0
Molecular Formula C8H19NO
Molecular Weight (g/mol) 145.246
MDL Number MFCD02094315
InChI Key WDCOJSGXSPGNFK-UHFFFAOYSA-N
PubChem CID 15919832
IUPAC Name 8-aminooctan-1-ol
SMILES C(CCCCO)CCCN

Specifications

Melting Point 65°C
Color White-Yellow
Quantity 1 g
Formula Weight 145.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Amino-1-octanol