missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™ Laboratory Detergents

Catalog No. icn7667093
Change view
Click to view available options
Volume (English):
1 gal.
5 gal.
4 x 1 gal.
55 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
ICN7667093 1 gal.
ICN7667094 4 x 1 gal.
ICN7667095 5 gal.
ICN7667098 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN7667093 Supplier MP Biomedicals™ Supplier No. 097667093

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Safe, effective cleaning solutions for glass and plastic labware

7X cleaning solution was developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments

  • Effective and safe
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration

Specifications

Form Liquid
pH 6 to 7.5
Concentration 7X
Content And Storage 15°C to 30°C
For Use With (Application) Use on Glass, Plastic, benchtop equipment and tubing
Volume (English) 1 gal.