missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X Cleaning Solution

Catalog No. MP0976670A4
Change view
Click to view available options
Volume (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
MP0976670A4 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0976670A4 Supplier MP Biomedicals™ Supplier No. 0976670A4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Case of 12

Clean test tubes, petri-dishes, flasks, cylinders, burettes, pipettes, slide cover glasses, hypodermic syringes and all other types of laboratory glassware and surgical instruments. 7X Cleaning Solution is ideal for use whenever a high degree of cleanliness is required.

Applications: 7X cleaning solution was developed for use whenever a high degree of cleanliness is required. Test tubes, petri-dishes, flasks, cylinders, burettes, pipettes, slide cover glasses, hypodermic syringes and all other types of laboratory glassware and surgical instruments can be cleaned with 7X solution. Whenever critical cleaning procedures are involved, the use of 7X solution is indicated. 7X detergents show a broad range of application other than glassware, and have been used to clean re-usable parts of the endoscopic instruments, needles, laboratory surfaces, and stainless stain instruments and titanium components without damaging the support material. Moreover, optical components of instruments are efficiently cleaned because no fluorescent residues are left after rinsing.
Product Overview: 7X solutions are immediately and completely soluble in water at any concentration, without agitation. With normal detergents the precipitation of solids on cleaned surfaces is a major problem. The powerful dispersing agents in the 7X solutions work to reduce agglomerates to individual particles and hold them in suspension so that they are not redeposited on the surfaces being cleaned. 7X solutions contain a powerful wetting agent which will penetrate and clean minute crevices not accessible to ordinary cleaners. Laboratory equipment and glassware washed in any of the 7X solutions drain completely in a few seconds. Solutions which do not drain quickly can allow molecules of solute to be deposited on the washed surface to the extent that a visible film is present. Such deposits can affect critical experimental work. 7X cleaning solutions are compatible with alkalis and acids. The pH is near normal and varies only minimally with the concentration. The neutral pH levels will not induce etching of delicate glassware, thereby reducing weight loss and premature breakage. 7X cleaning solutions are of particular value in cell culture laboratories where any trace of chemical contamination can interfere with the growth of cell cultures in tubes or flasks.
Soluble in water at ANY concentration
Safe for glass or plastic labware

Specifications

Form Cloudy Liquid
pH 6.0 to 7.5
Appearance Cloudy Blue
Composition 7X Cleaning Solutions are a concentrated liquid, made up of anionic surface active agent and special
Content And Storage Store at Room Temperature (15°C to 30°C), All 7X Cleaning Solutions have an indefinite shelf life in their original containers, at room temperature.
Description Lab Essentials, 7X is completely soluble in water at any concentration.
Quantity Case of 12
Volume (English) 16 oz.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.