missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 7900HT TaqMan™ Low Density Array Instrument Verification RNase P Kit

Catalog No. 4351468
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4351468 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4351468 Supplier Applied Biosystems™ Supplier No. 4351468
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used to validate the performance of the Applied Biosystems™ 7900HT Fast Real-Time PCR Systems with the TaqMan™ Low Density Array Block Upgrade.

The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each vial contains premixed reagents (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.

Specifications

Content And Storage Contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents.
For Use With (Equipment) 7900HT System
Product Type Spectral Calibration Kit
For Use With (Application) Real time PCR (qPCR)
Description Fluorescent Detection
Quantity 1 Kit
Calibration Type Instrument Verification Plate, RNase P Calibration