Learn More
Applied Biosystems™ 7900HT System Standard Upgrade Kits
Description
The interchangeable 96- and 384-well block capability combined with the optional Automation Accessory offer unmatched flexibility. These features make the 7900HT system ideally suited to meet the high-throughput requirements of today’s drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.
4331405, 96-well block
Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
4331406, 384-well block
Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions
Spécifications
Spécifications
| For Use With (Equipment) | 7900HT System |
| Product Type | Block Upgrade Kit |
| For Use With (Application) | Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling |
| Description | 384-well Kit |
| Quantity | 1 Kit |