Learn More
Applied Biosystems™ 7900HT System Standard Upgrade Kits
Catalog No. 4331405
Encompass
Change view
Click to view available options
Description:
384-well Kit
96-well Kit
Quantity:
1 Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
4331405 96-well Kit 1 Kit
4331406 384-well Kit 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4331405 Supplier Applied Biosystems™ Supplier No. 4331405
Only null left
Optional accessory for the 7900HT Fast RT-PCR System, which is available is either a 96- or 384-well block format.
The interchangeable 96- and 384-well block capability combined with the optional Automation Accessory offer unmatched flexibility. These features make the 7900HT system ideally suited to meet the high-throughput requirements of today’s drug discovery process. Key applications include gene expression quantitation and the detection of single nucleotide polymorphisms (SNPs) using the fluorogenic 5' nuclease assay.The 7900HT Fast Real-Time PCR System is the only real-time quantitative PCR system that combines 96- and 384-well plate compatibility and the TaqMan™ Low Density Array with fully automated robotic loading-and now also offers optional Fast real-time PCR capability.
4331405, 96-well block
Easy-to-use and provides accurate discrimination of the fluorescence signal of individual dyes in the reaction mixture
4331406, 384-well block
Includes the Passive Reference I dye standard for signal normalization in all real-time and endpoint reactions
Specifications
For Use With (Equipment) 7900HT System
Product Type Block Upgrade Kit
For Use With (Application) Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling
Description 96-well Kit
Quantity 1 Kit