missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 7712 Series PVC Coated Gloves

Catalog No. 2870507
Change view
Click to view available options
Grip Design:
Rough
Smooth
Series:
7712 Series
7712R Series
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grip Design Series
2870507 Smooth 7712 Series
1139515 Rough 7712R Series
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2870507 Supplier SHOWA™ Supplier No. 771210

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PVC coated, multi-purpose gloves with soft cotton liner are designed for all industrial applications and protect against a range of hazards including chemicals, oils and abrasion.

Both styles are ideal for working with chemicals, oil, or grease in refining, mining, automotive subassembly, or construction

  • Fully coated PVC with smooth or rough finish
  • Cotton jersey lined Sanitized™
  • Recyclable
  • Washable

Specifications

Certifications/Compliance CFIA
Color Black
Latex Free Yes
Material PVC
Size Large
For Use With (Application) Chemical spray and treatment, coating preparation, laboratory, pharma and analysis, washing and cleaning
Glove Cuff Style Gauntlet
Grip Design Smooth
Liner Material Cotton
Lining Lined
Model 7712-10
Series 7712 Series
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less