Learn More
7500 Fast System Upgrade Kit
Description
The Applied Biosystems™ 7500 Fast Real-Time PCR System enables standard 96-well format high speed thermal cycling, significantly reducing your run time for quantitative real-time PCR applications, delivering results in about 30 minutes. The Upgrade Kit is available to upgrade a standard 7500 Real-Time PCR System to the Fast Configuration via a field service installation.
Specifications
Specifications
| Content And Storage | Package includes 7500 Fast Block replacement and supporting system updates. Recommended operating conditions; Temperature: 15 to 30 °C (59 to 86 °F). Maximum change of less than 15 degrees Celsius (27 °F) per 24 hrs. Humidity: 20 to 80% relative humidity, noncondensing. |
| For Use With (Equipment) | 7500 Fast System, 7500 System |
| Product Type | System Upgrade Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Quantity | 1 kit |
Frequently Asked Questions (FAQs)
For the 7500 Fast real time PCR systems, the thermal cycling parameters are defaulted to 50°C/2 min for UNG incubation, 95°C/20 sec for fast polymerase activation, 40 cycles of 95°C/3 secs for the dissociation and 60°C/30 seconds for annealing and extensions.
Yes. you can choose either standard or fast ramp on the 7500 Fast System for software versions 2x or greater, and a 9600 emulation thermal cycling mode is available for customers who wish to run assays based on adjusted ramp rate. However, you should be aware that we cannot guarantee identical performance to the same modes run on a standard 7500 system block.
Runs can be compared between standard and fast mode, as the data collected are collected in all five filters in the instrument. However, keep in mind that these data may not always show identical results and may have added variability due to the data collection processes and differences in ramp rate between standard and fast mode.
Yes, older 7500 or 7500 fast data files are compatible with the current version of the SDS software. However, there is one exception: please note that the SDS v1.3 software will not allow RQ Studies to be created from RQ Plates from different instrument types, different thermal modes (i.e., fast and standard modes) or different numbers of thermal cycles.
You can use up to Windows XP, Service Pack 3 (SP3) on the computer that is connected to the 7500 Fast Real-Time PCR System instrument. An exception is the SDS v2.0.5 software, which can be used with the Windows 7 operating system for stand-alone data analysis steps.
For Research Use Only. Not for use in diagnostic procedures.