missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 7500 Fast Real-Time PCR Systems Spectral Calibration Kit II

Catalog No. 4362201
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4362201 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4362201 Supplier Applied Biosystems™ Supplier No. 4362201

Easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7500 Fast Real-Time PCR System.

This easy-to-use kit establishes the pure dye spectra and multicomponent values for Cy3™, Cy5™, and Texas Red™ dyes on the 7500 Fast Real-Time PCR System. The Applied Biosystems™ 7500 Real-Time PCR Systems Spectral Calibration Kit II contains three Optical 96-well Fast Thermocycling Plates preloaded and sealed with Cy3™, Cy5™, and Texas Red™ fluorescent dye standards. Spend Your Time on Research, Not Instrument Calibration: This kit's easy-to-use, preloaded, 96-well optical reaction plates enable quick and easy calibration of the 7500 Fast Real-Time PCR System. Insert the reaction plates during system installation, and the system collects the dye standard's spectral information. The system's application algorithm then uses the calibration information to ensure accurate data analysis.

Kit II

  • 96-well optical reaction plates preloaded with Cy3™, Cy5™, and Texas Red™ dyes allow quick and easy calibration of the 7500 Fast Real-Time PCR System
  • Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously

Kit l

  • 96-well optical reaction plates preloaded with dyes (FAM™/SYBR™ Green I, VIC™/JOE™, NED™/TAMRA™/ROX™) allow quick and easy calibration of the 7500 Fast Real-Time PCR System
  • Easy-to-use, preloaded spectral calibration kit allows accurate discrimination of the individual dyes in a reaction mixture when all the dyes are excited simultaneously
  • Includes the Passive Reference I dye standard required for signal normalization in all real-time and endpoint reactions

For Research Use Only. Not for use in diagnostic procedures.

Order Info

Expiration date: 12 months from the date of delivery.

Specifications

Content And Storage Contains three optical 96-Well Fast Thermocycling Plates preloaded and sealed, containing Cy3™ Texas Red™ and Cy5™ fluorescent dye standards.

Store at -15 to -25°C. Protect from light.

Expiration date: 12 months from the date of delivery
Calibrated Dye Cy™3, Cy™5, Texas Red™
For Use With (Equipment) 7500 Fast System
Detection Method Fluorescence
Product Type Spectral Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 kit
Calibration Type Spectral Calibration

For Research Use Only. Not for use in diagnostic procedures.