missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 7166 Nitrile Coated Oil & Grease Resistant Gloves

Catalog No. 29632801
Change view
Click to view available options
Exterior Finish:
Fully Textured
No Texture
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Exterior Finish
29632801 Fully Textured
296354723 No Texture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29632801 Supplier SHOWA™ Supplier No. 7166R10
Only null left

Full nitrile coating is impermeable to oils and chemicals, while also protecting against abrasions and punctures. Soft cotton liner is soft, comfortable, wicks away sweat, and controls temperature.

  • Exclusive coating provides cut, puncture, snag, and abrasion resistance
  • Smooth finish enhances grip in dry and oily conditions
  • Rough coating enhances grip and adds durability
  • Barrier is resistant to oil and grease, and prevents oil breakthrough
  • Reinforced safety cuff for additional protection
  • Economical replacement for leather

Specifications

Construction Cut and Sewn
Cuff Material Reinforced Cotton
Coating Material Nitrile
Color Black
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Chemical Spray/Treatment, Landscaping, and Utility Work
Glove Cuff Color Natural
Glove Cuff Style Safety
Series 7166R
Show More Show Less