Learn More
Excelta™ 7000 Series Precision Wire Cutters With Angled Head
Features set-screw to prolong cutting life and prevent over-travel of cutting blades
Supplier:  Excelta™ 7204E
 View more versions of this product
Catalog No. 17467441
Only null left
Description
Description
  • Maximum flush cut
  • Set-screw Stop prevents premature wear
  • Induction hardened carbon tool steel
Specifications
Chemical Identifiers
Ultra Precision
Carbon Tool Steel
Maximum Flush
Specifications
Standard
Large Angulated Tapered
13.3 cm
0.003 to 0.025 in.
Molded Grips
0.5 in.
1.27 cm
0.25 in.
Ultra Precision
5.25 in.
Carbon Tool Steel
Maximum Flush
Set-screw, 40° angled tip with 0.3 in. induction hardened cutting blades to RHC 63-65 and dual leaf springs provide smooth action
0.3 in.
0.76 cm
0.63 cm
Product Suggestions
Product Suggestions