missing translation for 'onlineSavingsMsg'
Learn More

Excelta™ 7000 Series Precision Wire Cutters With Angled Head

Features set-screw to prolong cutting life and prevent over-travel of cutting blades

Supplier:  Excelta™ 7104E

 View more versions of this product

Catalog No. 17467412


Only null left

Description

Description

  • Maximum flush cut
  • Set-screw Stop prevents premature wear
  • Induction hardened carbon tool steel
Specifications

Chemical Identifiers

Ultra Precision
Carbon Tool Steel
Maximum Flush

Specifications

Ergonomic long
Angulated Tapered
15.24 cm
0.003 to 0.25 in.
Long Grips
0.5 in.
1.27 cm
0.025 in.
Ultra Precision
6 in.
Carbon Tool Steel
Maximum Flush
Set-screw, 40° angled tip with 0.3 in. cutting blades and dual leaf springs provide smooth action
0.3 in.
0.76 cm
0.063 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions