Learn More
7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%
CAS: 450-62-4 | C10H10F3N | 201.192 g/mol
Supplier: Thermo Scientific Chemicals L1015006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline | |
| 30°C to 34°C | |
| C10H10F3N | |
| 5 g | |
| 7-trifluoromethyl-1,2,3,4-tetrahydroquinoline, 7-trifluoromethyl-1,2,3,4-tetrahydro-quinoline, quinoline, 1,2,3,4-tetrahydro-7-trifluoromethyl, pubchem7221, acmc-20aip4 | |
| C1CC2=C(C=C(C=C2)C(F)(F)F)NC1 | |
| 201.192 | |
| 201.19 |
| 450-62-4 | |
| 135°C to 140°C (22 mmHg) | |
| MFCD00079784 | |
| 153990 | |
| RGZZKZNESVFQKR-UHFFFAOYSA-N | |
| 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline | |
| 2781139 | |
| 97% |
Chemical Identifiers
| 450-62-4 | |
| 201.192 | |
| RGZZKZNESVFQKR-UHFFFAOYSA-N | |
| 2781139 | |
| C1CC2=C(C=C(C=C2)C(F)(F)F)NC1 |
| C10H10F3N | |
| MFCD00079784 | |
| 7-trifluoromethyl-1,2,3,4-tetrahydroquinoline, 7-trifluoromethyl-1,2,3,4-tetrahydro-quinoline, quinoline, 1,2,3,4-tetrahydro-7-trifluoromethyl, pubchem7221, acmc-20aip4 | |
| 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only