missing translation for 'onlineSavingsMsg'
Learn More

7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%

Catalog No. AAL10150MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL10150MD 250 mg
AAL1015003 1 g
AAL1015006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL10150MD Supplier Thermo Scientific Chemicals Supplier No. L10150MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 450-62-4 | C10H10F3N | 201.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 450-62-4
Molecular Formula C10H10F3N
Molecular Weight (g/mol) 201.192
MDL Number MFCD00079784
InChI Key RGZZKZNESVFQKR-UHFFFAOYSA-N
Synonym 7-trifluoromethyl-1,2,3,4-tetrahydroquinoline, 7-trifluoromethyl-1,2,3,4-tetrahydro-quinoline, quinoline, 1,2,3,4-tetrahydro-7-trifluoromethyl, pubchem7221, acmc-20aip4
PubChem CID 2781139
IUPAC Name 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline
SMILES C1CC2=C(C=C(C=C2)C(F)(F)F)NC1

Specifications

Melting Point 30°C to 34°C
Boiling Point 135°C to 140°C (22 mmHg)
Quantity 250 mg
Beilstein 153990
Formula Weight 201.19
Percent Purity 97%
Chemical Name or Material 7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only