missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Tridecanone 97.0+%, TCI America™
$109.98 - $436.19
Chemical Identifiers
| CAS | 462-18-0 |
|---|---|
| Molecular Formula | C13H26O |
| Molecular Weight (g/mol) | 198.35 |
| MDL Number | MFCD00009543 |
| InChI Key | ULIAPOFMBCCSPE-UHFFFAOYSA-N |
| Synonym | 7-tridecanone, dihexyl ketone, enanthone, hexyl ketone, di-n-hexyl ketone, dihexadecyl ketone, unii-l0rdd0t8dg, l0rdd0t8dg, dihexylketone, di-normal-hexyl ketone |
| PubChem CID | 10015 |
| IUPAC Name | tridecan-7-one |
| SMILES | CCCCCCC(=O)CCCCCC |
Chemical Identifiers
| 462-18-0 | |
| 198.35 | |
| ULIAPOFMBCCSPE-UHFFFAOYSA-N | |
| 10015 | |
| CCCCCCC(=O)CCCCCC |
| C13H26O | |
| MFCD00009543 | |
| 7-tridecanone, dihexyl ketone, enanthone, hexyl ketone, di-n-hexyl ketone, dihexadecyl ketone, unii-l0rdd0t8dg, l0rdd0t8dg, dihexylketone, di-normal-hexyl ketone | |
| tridecan-7-one |
Specifications
| 462-18-0 | |
| 264°C | |
| MFCD00009543 | |
| 7-tridecanone, dihexyl ketone, enanthone, hexyl ketone, di-n-hexyl ketone, dihexadecyl ketone, unii-l0rdd0t8dg, l0rdd0t8dg, dihexylketone, di-normal-hexyl ketone | |
| CCCCCCC(=O)CCCCCC | |
| 198.35 | |
| 198.35 | |
| Crystalline Lumps |
| White-Yellow | |
| C13H26O | |
| 5 g | |
| ULIAPOFMBCCSPE-UHFFFAOYSA-N | |
| tridecan-7-one | |
| 10015 | |
| ≥97.0% (GC) | |
| 7-Tridecanone |
Safety and Handling
EINECSNumber : (2)-2475
RTECSNumber : YD4577000
TSCA : No