Learn More
Eisco™ 7 Piece Dissecting Set
Premium quality stainless-steel dissecting set in a beautiful leather case.
Supplier:  Eisco™ BI0152
Catalog No. S14005
Only null left
Description
Description
  • (2) Scissors
  • (2) Forceps
  • (1) Probe and seeker
  • (1) Scalpel handle with sterile blade
  • (1) Straight needle with wooden handle
Specifications
Specifications
Dissection Set
Reusable
2 scissors (4 in. fine point, 5 in. blunt end), 2 forceps (4 in. fine point, 5 in. blunt end), 1 seeker with blunt end needle, 1 straight needle with 4 in. wooden handle, 1 scalpel handle #4, 1 scalpel blade #20
7 in.
0.4 lb.
7 piece kit in zipper case
For basic dissection lessons
7 pieces
17.78 cm
Non-sterile
Product Suggestions
Product Suggestions