missing translation for 'onlineSavingsMsg'
Learn More

7-Phenylheptanoic acid, 97%

CAS: 40228-90-8 | C13H18O2 | 206.285 g/mol

Supplier:  Thermo Scientific Chemicals L0500906

 View more versions of this product

Catalog No. AAL0500906


Only null left
Explore available promotions

Description

Description

7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Specifications

7-Phenylheptanoic acid
∼20°C
147°C to 150°C (0.4 mmHg)
C13H18O2
MFCD00037801
1957081
Insoluble in water.
C1=CC=C(C=C1)CCCCCCC(=O)O
206.285
206.29
40228-90-8
1.009
>110°C (230°F)
1.507
5 g
benzeneheptanoic acid, phenylheptanoic acid, 7-phenyl-heptanoic acid, acmc-20alwn, 7-phenylheptanoic acid, xxc
ZVSXKFNTWOIGJI-UHFFFAOYSA-N
7-phenylheptanoic acid
520987
97%

Chemical Identifiers

40228-90-8
206.285
ZVSXKFNTWOIGJI-UHFFFAOYSA-N
520987
C1=CC=C(C=C1)CCCCCCC(=O)O
C13H18O2
MFCD00037801
benzeneheptanoic acid, phenylheptanoic acid, 7-phenyl-heptanoic acid, acmc-20alwn, 7-phenylheptanoic acid, xxc
7-phenylheptanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only