missing translation for 'onlineSavingsMsg'
Learn More

7-Phenylheptanoic acid, 97%

Catalog No. AAL0500903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0500903 1 g
AAL0500906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0500903 Supplier Thermo Scientific Chemicals Supplier No. L0500903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40228-90-8 | C13H18O2 | 206.285 g/mol

7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Phenylheptanoic acid is used in the process to make amino acid derivatives.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 40228-90-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00037801
InChI Key ZVSXKFNTWOIGJI-UHFFFAOYSA-N
Synonym benzeneheptanoic acid, phenylheptanoic acid, 7-phenyl-heptanoic acid, acmc-20alwn, 7-phenylheptanoic acid, xxc
PubChem CID 520987
IUPAC Name 7-phenylheptanoic acid
SMILES C1=CC=C(C=C1)CCCCCCC(=O)O

Specifications

Melting Point ∼20°C
Density 1.009
Boiling Point 147°C to 150°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.507
Quantity 1 g
Beilstein 1957081
Solubility Information Insoluble in water.
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material 7-Phenylheptanoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only