missing translation for 'onlineSavingsMsg'
Learn More

7-Oxabicyclo[2.2.1]heptane, 98%

Catalog No. p-7026557
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2261206 5 g
AAB2261214 25 g
AAB2261222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2261206 Supplier Thermo Scientific Chemicals Supplier No. B2261206
Only null left

CAS: 279-49-2 | C6H10O | 98.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 279-49-2
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00082342
InChI Key YPWFNLSXQIGJCK-UHFFFAOYSA-N
Synonym 1,4-epoxycyclohexane, 7-oxabicyclo 2.2.1 heptane, 7-oxanorbornane, 1,4-oxycyclohexane, 3,6-endoxocyclohexane, 3,6-endooxycyclohexane, cyclohexane,4-epoxy, cyclohexane,6-endoxo, cyclohexane, 1,4-epoxy
PubChem CID 9234
IUPAC Name 7-oxabicyclo[2.2.1]heptane
SMILES C1CC2CCC1O2

Specifications

Density 0.968
Quantity 5 g
Refractive Index 1.448
UN Number UN3271
Boiling Point 118°C to 120°C (713 mmHg)
Formula Weight 98.15
Percent Purity 98%
Chemical Name or Material 7-Oxabicyclo[2.2.1]heptane
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 205-998-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only