missing translation for 'onlineSavingsMsg'
Learn More

7-Octenyl Acetate 98.0+%, TCI America™

Catalog No. p-7117477
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15015G 5 g
A150125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15015G Supplier TCI America Supplier No. A15015G

Chemical Identifiers

CAS 5048-35-1
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD01861278
InChI Key DBRVFNFZYZFKRC-UHFFFAOYSA-N
Synonym 8-Acetoxy-1-octene, Acetic Acid 7-Octenyl Ester
PubChem CID 13990525
IUPAC Name oct-7-en-1-yl acetate
SMILES CC(=O)OCCCCCCC=C

Specifications

Color Colorless
Boiling Point 104°C
Quantity 5 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7-Octenyl Acetate
EINECSNumber (2)-0760
TSCA No