missing translation for 'onlineSavingsMsg'
Learn More

7-Nitroindazole 97.0+%, TCI America™

Catalog No. N08271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08271G 1 g
N08275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08271G Supplier TCI America Supplier No. N08271G

Chemical Identifiers

CAS 2942-42-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00022789
InChI Key PQCAUHUKTBHUSA-UHFFFAOYSA-N
Synonym 7-nitroindazole, 1h-indazole, 7-nitro, 7-ni, 7-nitro-indazole, unii-ux0n37cmvh, ccris 3309, 2h-indazole,7-nitro, ux0n37cmvh, 7ni
PubChem CID 1893
IUPAC Name 7-nitro-1H-indazole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2

Specifications

Melting Point 186°C
Quantity 1 g
Formula Weight 163.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Nitroindazole
RTECSNumber NK7962200
TSCA No