missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Catalog No. p-7041099
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61048MD 250 mg
AAH6104803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH61048MD Supplier Thermo Scientific Chemicals Supplier No. H61048MD
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69373-37-1
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.22
MDL Number MFCD23379852
InChI Key LCOAYUPAESYAHF-UHFFFAOYSA-N
Synonym 7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 20496121
IUPAC Name 7-nitro-3,4-dihydro-2H-1,4-benzothiazine
SMILES [O-][N+](=O)C1=CC2=C(NCCS2)C=C1

Specifications

Quantity 250 mg
Formula Weight 196.22
Percent Purity 97%
Chemical Name or Material 7-Nitro-3,4-dihydro-2H-1,4-benzothiazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only