Learn More
7-Nitro-1H-indazole, 98%
CAS: 2942-42-9 | C7H5N3O2 | 163.136 g/mol
Supplier: Thermo Scientific Chemicals L0797006
Description
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 7-Nitro-1H-indazole | |
| 184°C to 189°C | |
| MFCD00022789 | |
| 6809 | |
| Insoluble in water. | |
| C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2 | |
| 163.136 | |
| 163.14 |
| 2942-42-9 | |
| C7H5N3O2 | |
| 5 g | |
| 7-nitroindazole, 1h-indazole, 7-nitro, 7-ni, 7-nitro-indazole, unii-ux0n37cmvh, ccris 3309, 2h-indazole,7-nitro, ux0n37cmvh, 7ni | |
| PQCAUHUKTBHUSA-UHFFFAOYSA-N | |
| 7-nitro-1H-indazole | |
| 1893 | |
| 98% |
Chemical Identifiers
| 2942-42-9 | |
| 163.136 | |
| PQCAUHUKTBHUSA-UHFFFAOYSA-N | |
| 1893 | |
| C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2 |
| C7H5N3O2 | |
| MFCD00022789 | |
| 7-nitroindazole, 1h-indazole, 7-nitro, 7-ni, 7-nitro-indazole, unii-ux0n37cmvh, ccris 3309, 2h-indazole,7-nitro, ux0n37cmvh, 7ni | |
| 7-nitro-1H-indazole |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H319-H341
EINECSNumber : 220-934-4
RTECSNumber : NK7962200
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only