missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1H-indazole, 98%

Catalog No. AAL0797003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0797003 1 g
AAL0797006 5 g
AAL0797014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0797003 Supplier Thermo Scientific Chemicals Supplier No. L0797003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2942-42-9 | C7H5N3O2 | 163.136 g/mol

7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-nitroindazole is a specific inhibitor of neural nitric oxide synthase (nNOS), an enzyme that converts arginine to citrulline and nitric oxide (NO) in neuronal tissue. NO is a key player in the hippocampal long-term potentiation (LTP) and in general cell signaling.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 2942-42-9
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00022789
InChI Key PQCAUHUKTBHUSA-UHFFFAOYSA-N
Synonym 7-nitroindazole, 1h-indazole, 7-nitro, 7-ni, 7-nitro-indazole, unii-ux0n37cmvh, ccris 3309, 2h-indazole,7-nitro, ux0n37cmvh, 7ni
PubChem CID 1893
IUPAC Name 7-nitro-1H-indazole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NN=C2

Specifications

Melting Point 184°C to 189°C
Quantity 1 g
Beilstein 6809
Solubility Information Insoluble in water.
Formula Weight 163.14
Percent Purity 98%
Chemical Name or Material 7-Nitro-1H-indazole
Hazard Category H302+H312+H332-H319-H341
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P501c
EINECSNumber 220-934-4
RTECSNumber NK7962200
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only