missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Nitro-1,2,3,4-tetrahydroquinoline 98.0+%, TCI America™
Supplier: TCI America N095525G
Specifications
| 7-Nitro-1,2,3,4-tetrahydroquinoline | |
| 63°C | |
| MFCD00496654 | |
| 1,2,3,4-tetrahydro-7-nitroquinoline, 7-nitro-1,2,3,4-tetrahydro quinoline, quinoline, 1,2,3,4-tetrahydro-7-nitro, 7-nitro-1,2,3,4-tetrahydro-quinoline, pubchem7584, acmc-1aglc, 7-nitrotetrahydro-quinoline, 7-nitro-1,2,3,4-terahydroquinoline | |
| C1CC2=C(C=C(C=C2)[N+](=O)[O-])NC1 | |
| 178.191 | |
| 178.19 | |
| Crystalline Powder |
| 30450-62-5 | |
| C9H10N2O2 | |
| 25 g | |
| WSWMGHRLUYADNA-UHFFFAOYSA-N | |
| 7-nitro-1,2,3,4-tetrahydroquinoline | |
| 252513 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 30450-62-5 | |
| 178.191 | |
| WSWMGHRLUYADNA-UHFFFAOYSA-N | |
| 252513 | |
| C1CC2=C(C=C(C=C2)[N+](=O)[O-])NC1 |
| C9H10N2O2 | |
| MFCD00496654 | |
| 1,2,3,4-tetrahydro-7-nitroquinoline, 7-nitro-1,2,3,4-tetrahydro quinoline, quinoline, 1,2,3,4-tetrahydro-7-nitro, 7-nitro-1,2,3,4-tetrahydro-quinoline, pubchem7584, acmc-1aglc, 7-nitrotetrahydro-quinoline, 7-nitro-1,2,3,4-terahydroquinoline | |
| 7-nitro-1,2,3,4-tetrahydroquinoline |
Safety and Handling
TSCA : No