missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1,2,3,4-tetrahydroquinoline 98.0+%, TCI America™

Catalog No. N09555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09555G 5 g
N095525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09555G Supplier TCI America Supplier No. N09555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30450-62-5
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.191
MDL Number MFCD00496654
InChI Key WSWMGHRLUYADNA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-7-nitroquinoline, 7-nitro-1,2,3,4-tetrahydro quinoline, quinoline, 1,2,3,4-tetrahydro-7-nitro, 7-nitro-1,2,3,4-tetrahydro-quinoline, pubchem7584, acmc-1aglc, 7-nitrotetrahydro-quinoline, 7-nitro-1,2,3,4-terahydroquinoline
PubChem CID 252513
IUPAC Name 7-nitro-1,2,3,4-tetrahydroquinoline
SMILES C1CC2=C(C=C(C=C2)[N+](=O)[O-])NC1

Specifications

Melting Point 63°C
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Nitro-1,2,3,4-tetrahydroquinoline