missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1,2,3,4-tetrahydroquinoline, 95%

Catalog No. p-7053871
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6161203 1 g
AAH6161206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6161203 Supplier Thermo Scientific Chemicals Supplier No. H6161203
Only null left

CAS: 30450-62-5 | C9H10N2O2 | 178.191 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30450-62-5
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.191
MDL Number MFCD00496654
InChI Key WSWMGHRLUYADNA-UHFFFAOYSA-N
Synonym 1,2,3,4-tetrahydro-7-nitroquinoline, 7-nitro-1,2,3,4-tetrahydro quinoline, quinoline, 1,2,3,4-tetrahydro-7-nitro, 7-nitro-1,2,3,4-tetrahydro-quinoline, pubchem7584, acmc-1aglc, 7-nitrotetrahydro-quinoline, 7-nitro-1,2,3,4-terahydroquinoline
PubChem CID 252513
IUPAC Name 7-nitro-1,2,3,4-tetrahydroquinoline
SMILES C1CC2=C(C=C(C=C2)[N+](=O)[O-])NC1

Specifications

Color Red
Quantity 1 g
Formula Weight 178.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 7-Nitro-1,2,3,4-tetrahydroquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only