missing translation for 'onlineSavingsMsg'
Learn More

7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97+%

Catalog No. AAH52389MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52389MD 250 mg
AAH5238903 1 g
AAH5238906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52389MD Supplier Thermo Scientific Chemicals Supplier No. H52389MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99365-69-2 | C9H11ClN2O2 | 214.649 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99365-69-2
Molecular Formula C9H11ClN2O2
Molecular Weight (g/mol) 214.649
MDL Number MFCD07371494
InChI Key KGIXLJRTYZOUCW-UHFFFAOYSA-N
Synonym 7-nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydro-isoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline hcl, 1,2,3,4-tetrahydro-7-nitroisoquinoline hydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 1,2,3,4-tetrahydro-7-nitro-, monohydrochloride, ksc683k9f, 7-nitro-1,2,3,4-tetrahydroisoquinoline monohydrochloride, 7-nitro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 13521670
IUPAC Name 7-nitro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)[N+](=O)[O-].Cl

Specifications

Quantity 250 mg
Formula Weight 214.65
Percent Purity ≥97%
Chemical Name or Material 7-Nitro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only