missing translation for 'onlineSavingsMsg'
Learn More

7-Methylxanthine, 98%

Catalog No. AAH29190MF
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH29190MF 50 mg
AAH29190MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH29190MF Supplier Thermo Scientific Chemicals Supplier No. H29190MF
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 552-62-5 | C6H6N4O2 | 166.14 g/mol

It is used to metabolite theophylline and caffeine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to metabolite theophylline and caffeine.

Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 552-62-5
Molecular Formula C6H6N4O2
Molecular Weight (g/mol) 166.14
MDL Number MFCD00037979
InChI Key PFWLFWPASULGAN-UHFFFAOYSA-N
Synonym 7-methylxanthine, heteroxanthine, heteroxanthin, 7-methylxanthin, 7-methyl-1h-purine-2,6 3h,7h-dione, 2,6-dihydroxy-7-methylpurine, xanthine, 7-methyl, 3,7-dihydro-7-methyl-1h-purine-2,6-dione, 7-methyl-3,7-dihydro-1h-purine-2,6-dione, 1h-purine-2,6-dione, 3,7-dihydro-7-methyl
PubChem CID 68374
ChEBI CHEBI:48991
SMILES CN1C=NC2=C1C(=O)NC(=O)N2

Specifications

Melting Point >300°C
Quantity 50 mg
Solubility Information Soluble in dimethyl sulfoxide. Insoluble in water.
IUPAC Name 7-methyl-3H-purine-2,6-dione
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 7-Methylxanthine
EINECSNumber 209-019-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only