missing translation for 'onlineSavingsMsg'
Learn More

7-Methylindole 98.0+%, TCI America™

Catalog No. M17025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17025G 5 g
M17021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17025G Supplier TCI America Supplier No. M17025G

Chemical Identifiers

CAS 933-67-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
InChI Key KGWPHCDTOLQQEP-UHFFFAOYSA-N
Synonym 7-methylindole, 7-methylindol, 1h-indole, 7-methyl, indole, 7-methyl, 7-methyindole, 7-methyl-indole, 7-methyl indole, unii-7z1e6hit9s, 7z1e6hit9s, 7-methyl-1h-indol-1-yl
PubChem CID 70275
IUPAC Name 7-methyl-1H-indole
SMILES CC1=CC=CC2=C1NC=C2

Specifications

Melting Point 83°C
Boiling Point 266°C
Quantity 5 g
Formula Weight 131.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methylindole