missing translation for 'onlineSavingsMsg'
Learn More

7-Methylindole, 98%

Catalog No. p-7047072
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAL1752103 1 g
AAL1752106 5 g
2 options

Catalog No. AAL1752103

Supplier: Thermo Scientific Chemicals L1752103

Only null left

CAS: 933-67-5 | C9H9N | 131.178 g/mol

It is a reactant for preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition, tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, plant growth inhibitors, antifungal agents, sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes and inhibitors of HIV-1 attachment. It is a reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for preparation of pharmaceutically active pyrrolidine analogues via Markovnikov addition, tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, plant growth inhibitors, antifungal agents, sodium-Dependent Glucose Co-transporter 2 (SGLT2) Inhibitors for the Management of Hyperglycemia in Diabetes and inhibitors of HIV-1 attachment. It is a reactant for Friedel-Crafts alkylation reactions and reductive formylation reactions.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 933-67-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005684
InChI Key KGWPHCDTOLQQEP-UHFFFAOYSA-N
Synonym 7-methylindole, 7-methylindol, 1h-indole, 7-methyl, indole, 7-methyl, 7-methyindole, 7-methyl-indole, 7-methyl indole, unii-7z1e6hit9s, 7z1e6hit9s, 7-methyl-1h-indol-1-yl
PubChem CID 70275
IUPAC Name 7-methyl-1H-indole
SMILES CC1=CC=CC2=C1NC=C2

Specifications

Melting Point 83°C to 84°C
Boiling Point 266°C
Quantity 1 g
Beilstein 111088
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 7-Methylindole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 213-270-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only