missing translation for 'onlineSavingsMsg'
Learn More

7-Methylindole-3-carboxaldehyde, 98%

Catalog No. AAL1751906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1751906 5 g
AAL1751903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1751906 Supplier Thermo Scientific Chemicals Supplier No. L1751906
Only null left

CAS: 4771-50-0 | C10H9NO | 159.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4771-50-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.19
MDL Number MFCD00047170
InChI Key KTUFZHVVJBHGKZ-UHFFFAOYSA-N
Synonym 7-methylindole-3-carboxaldehyde, 7-methylindole-3-carboxyaldehyde, 3-formyl-7-methylindole, 7-methylindole 3-carboxaldehyde, 1h-indole-3-carboxaldehyde, 7-methyl, 3-carboxaldehyde, 7-methylindole, 7-methyl-1h-indole-3-carboxaldehyde, 7-methylindole-3-carbaldehyde, pubchem8327, acmc-1ak55
PubChem CID 260389
IUPAC Name 7-methyl-1H-indole-3-carbaldehyde
SMILES CC1=C2NC=C(C=O)C2=CC=C1

Specifications

Melting Point 209°C to 211°C
Quantity 5 g
Beilstein 122340
Sensitivity Air sensitive
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 7-Methylindole-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only