missing translation for 'onlineSavingsMsg'
Learn More

7-Methylcoumarin 98.0+%, TCI America™

Catalog No. M123625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M123625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M123625G Supplier TCI America Supplier No. M123625G
Only null left

Chemical Identifiers

CAS 2445-83-2
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00006880
InChI Key DLHXRDUXNVEIEY-UHFFFAOYSA-N
Synonym 7-methylcoumarin, 2h-1-benzopyran-2-one, 7-methyl, 7-methyl-2h-chromen-2-one, coumarin, 7-methyl, 7-methyl coumarin, 7-methylcumarin, 7-methyl-2h-1-benzopyran-2-one, unii-ycn83t03qu, 7-methyl-coumarin, 7-methyl-chromen-2-one
PubChem CID 17131
IUPAC Name 7-methyl-2H-chromen-2-one
SMILES CC1=CC=C2C=CC(=O)OC2=C1

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 25 g
Formula Weight 160.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methylcoumarin
EINECSNumber (5)-0687
RTECSNumber GN7792100
TSCA Yes