missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-1,4-benzothiazin-3-one, TCI America™

Catalog No. M16611G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M16611G Supplier TCI America Supplier No. M16611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30322-02-2
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.24
MDL Number MFCD02660690
InChI Key ODJQMWDFNLNCTM-UHFFFAOYSA-N
PubChem CID 689104
IUPAC Name 7-methyl-3,4-dihydro-2H-1,4-benzothiazin-3-one
SMILES CC1=CC2=C(NC(=O)CS2)C=C1

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 1 g
Formula Weight 179.24
Physical Form Crystalline Powder
Chemical Name or Material 7-Methyl-1,4-benzothiazin-3-one